3-Chloro-2-formylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

14585-1G 14585-25G 14585-500MG 14585-5G
76907-638EA 142.17 USD
76907-638 76907-640 76907-642 76907-644
3-Chloro-2-formylphenylboronic acid ≥95%
3-Chloro-2-formylphenylboronic acid
Formula: C₇H₆BClO₃
MW: 184.385 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181639
CAS Number: 928048-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR