3-Bromo-5-methylpyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

14557-1G 14557-25G 14557-500MG 14557-5G
76907-622EA 260.99 USD
76907-622 76907-624 76907-626 76907-628
3-Bromo-5-methylpyridine-4-carboxaldehyde ≥97%
3-Bromo-5-methylpyridine-4-carboxaldehyde
Formula: C₇H₆BrNO
MW: 200.033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06410672
CAS Number: 203569-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR