3-Bromo-5-fluoro-2-propoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-fluoro-2-propoxyphenyl)boronic acid

14430-25G 14430-5G
76907-544EA 524.09 USD
76907-544 76907-546
3-Bromo-5-fluoro-2-propoxyphenylboronic acid ≥97%
3-Bromo-5-fluoro-2-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BBrFO₃
MW: 276.895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369707
CAS Number: 868272-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR