3-Chloro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14305-25G 14305-5G
76907-510EA 180.36 USD
76907-510 76907-512
3-Chloro-2-methoxybenzoic acid ≥97%
3-Chloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00179306
CAS Number: 3260-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR