(3-(Benzyloxy)-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-(Benzyloxy)-4-methylphenylboronic acid

14254-25G 14254-5G
76907-490EA 445.58 USD
76907-490 76907-492
(3-(Benzyloxy)-4-methylphenyl)boronic acid ≥97% research grade
(3-(Benzyloxy)-4-methylphenyl)boronic acid
Formula: C₁₄H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD11617255
CAS Number: 1256355-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR