3-Ethoxy-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

14249-1G 14249-25G 14249-500MG 14249-5G
76907-476EA 231.28 USD
76907-476 76907-478 76907-480 76907-482
3-Ethoxy-4-methylbenzaldehyde ≥95%
3-Ethoxy-4-methylbenzaldehyde
Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18375378
CAS Number: 157143-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR