3-Ethoxy-4-methylbenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14245-1G 14245-25G 14245-500MG 14245-5G
76907-466EA 153.84 USD
76907-466 76907-468 76907-470 76907-472
3-Ethoxy-4-methylbenzoic acid ≥97% research grade
3-Ethoxy-4-methylbenzoic acid
Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD14647108
CAS Number: 1250606-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR