3-Ethoxy-4-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14240-1G 14240-25G 14240-5G
76907-452EA 102.91 USD
76907-452 76907-454 76907-456
3-Ethoxy-4-methylphenylboronic acid ≥97% research grade
3-Ethoxy-4-methylphenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD18447586
CAS Number: 1451391-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR