3,6-Difluoro-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3,6-Difluoro-2-methoxyphenyl)boronic acid

14161-1G 14161-25G 14161-5G
76907-440EA 78.51 USD
76907-440 76907-442 76907-444
3,6-Difluoro-2-methoxyphenylboronic acid ≥97%
3,6-Difluoro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856009
CAS Number: 919355-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR