3-Formyl-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14117-1G 14117-25G 14117-500MG 14117-5G
76907-400EA 120.95 USD
76907-400 76907-402 76907-404 76907-406
3-Formyl-4-methylbenzoic acid ≥97%
3-Formyl-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.158 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20660432
CAS Number: 69526-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR