3-Ethyl-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14108-1G 14108-25G 14108-5G
76907-388EA 88.06 USD
76907-388 76907-390 76907-392
3-Ethyl-4-formylphenylboronic acid ≥97%
3-Ethyl-4-formylphenylboronic acid
Formula: C₉H₁₁BO₃
MW: 177.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617251
CAS Number: 1218790-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR