4-Bromo-1-chloro-2-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

14059-25G 14059-5G
76907-364EA 177.18 USD
76907-364 76907-366
4-Bromo-1-chloro-2-ethylbenzene ≥95%
4-Bromo-1-chloro-2-ethylbenzene
Formula: C₈H₈BrCl
Storage Temperature: Ambient
MDL Number: MFCD00672954
CAS Number: 289039-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR