3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Diethylcarbamoyl)-4-fluorophenylboronic acid, 5-Borono-N,N-diethyl-2-fluorobenzamide

14025-1G 14025-25G 14025-5G
76907-342EA 66.84 USD
76907-342 76907-344 76907-346
3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid ≥97%
3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MW: 239.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235047
CAS Number: 874219-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR