3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-(Ethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-(Ethylcarbamoyl)-4-fluorophenylboronic acid

14018-1G 14018-25G 14018-5G
76907-318EA 88.06 USD
76907-318 76907-320 76907-322
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid ≥97%
3-(Ethylcarbamoyl)-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 210.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235039
CAS Number: 874219-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR