4-Bromo-1-phenyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

13809-25G 13809-5G
76907-292EA 336.31 USD
76907-292 76907-294
4-Bromo-1-phenyl-1H-pyrazole ≥97%
4-Bromo-1-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00234505
CAS Number: 15115-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR