4-Bromo-1-isopropylpyrazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-isopropyl-1H-pyrazole

13798-25G 13798-5G
76907-286EA 233.4 USD
76907-286 76907-288
4-Bromo-1-isopropylpyrazole ≥97%
4-Bromo-1-isopropylpyrazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂
MW: 189.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801021
CAS Number: 313735-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR