4-Bromo-2,3,6-trifluorophenol ≥97%
Supplier: AOBChem USA
|
Warning
|
13735-1G
13735-25G
13735-500MG
13735-5G
76907-272EA
236.59
USD
76907-272
76907-274
76907-276
76907-278
4-Bromo-2,3,6-trifluorophenol ≥97%
4-Bromo-2,3,6-trifluorophenol
Formula:
C₆H₂BrF₃O MW: 226.979 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22414509 CAS Number: 192446-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...