4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(2-Carbamothioylhydrazinecarbonyl)phenylboronic acid

13615-1G 13615-25G 13615-5G
76907-254EA 63.66 USD
76907-254 76907-256 76907-258
4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid ≥97% research grade
4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid
Formula: C₈H₁₀BN₃O₃S
MW: 239.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027244
CAS Number: 957060-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR