4-Chloro-3,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Chloro-3,5-difluorophenyl)boronic acid, 3,5-Difluoro-4-chlorophenylboronic acid

13594-1G 13594-25G 13594-5G
76907-240EA 76.39 USD
76907-240 76907-242 76907-244
4-Chloro-3,5-difluorophenylboronic acid ≥97%
4-Chloro-3,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08706254
CAS Number: 864759-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR