4-(4-Chlorophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-((4-Chlorophenyl)carbamoyl)phenyl)boronic acid

13483-25G 13483-5G
76907-196EA 573.95 USD
76907-196 76907-198
4-(4-Chlorophenylcarbamoyl)phenylboronic acid ≥97%
4-(4-Chlorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027203
CAS Number: 874288-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR