4-(4-Bromophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-((4-Bromophenyl)carbamoyl)phenyl)boronic acid, 4-(4-Bromophenylcarbamoyl)benzeneboronic acid

13479-25G 13479-5G
76907-188EA 375.56 USD
76907-188 76907-190
4-(4-Bromophenylcarbamoyl)phenylboronic acid ≥97%
4-(4-Bromophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027201
CAS Number: 874287-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR