4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester, Piperidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

13465-25G 13465-5G
76907-176EA 240.83 USD
76907-176 76907-178
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester ≥97% research grade
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027083
CAS Number: 938043-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR