3-Chloro-2-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloropyridine-2-carboxaldehyde

13288-25G 13288-5G
76907-134EA 381.93 USD
76907-134 76907-136
3-Chloro-2-formylpyridine ≥97%
3-Chloro-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991855
CAS Number: 206181-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR