4-Amino-3-nitrophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amino-3-nitrophenylboronic Acid Pinacol Ester, 4-Amino-3-nitrophenylboronic acid, pinacol ester

13169-25G 13169-5G
76907-122EA 453.01 USD
76907-122 76907-124
4-Amino-3-nitrophenylboronic acid pinacol ester ≥97%
4-Amino-3-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06795680
CAS Number: 833486-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR