4-Amino-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

13166-25G 13166-5G
76907-114EA 453.01 USD
76907-114 76907-116
4-Amino-2-fluorophenylboronic acid pinacol ester ≥97%
4-Amino-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD09951877
CAS Number: 819057-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR