4-Amino-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amino-2-methylphenylboronic acid, pinacol ester, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

13138-1G 13138-25G 13138-500MG 13138-5G
76907-100EA 171.87 USD
76907-100 76907-102 76907-104 76907-106
4-Amino-2-methylphenylboronic acid pinacol ester ≥97%
4-Amino-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458198
CAS Number: 631911-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR