3-Bromo-2,6-difluoropyridine ≥97%

Supplier: AOBChem USA

Warning

13087-25G 13087-5G
76907-088EA 289.63 USD
76907-088 76907-090
3-Bromo-2,6-difluoropyridine ≥97%
3-Bromo-2,6-difluoropyridine
Formula: C₅H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD11044274
CAS Number: 80392-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR