tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-(tert-Butoxycarbonylamino)-3-fluorophenylboronic acid, pinacol ester
13052-1G
13052-25G
13052-5G
76907-078EA
79.57
USD
76907-078
76907-080
76907-082
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate ≥95%
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate
Precaución: for Research Use Only
Formula:
C₁₇H₂₅BFNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD09033883 CAS Number: 262444-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...