3-Fluoro-4-(propan-2-yloxy)phenol ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-isopropoxyphenol

12962-1G 12962-25G 12962-500MG 12962-5G
76907-064EA 168.69 USD
76907-064 76907-066 76907-068 76907-070
3-Fluoro-4-(propan-2-yloxy)phenol ≥95% research grade
3-Fluoro-4-(propan-2-yloxy)phenol
Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16997390
CAS Number: 1243280-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR