3-(Propylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N-Propylaminocarbonyl)benzeneboronic acid, 3-(N-Propylaminocarbonyl)phenylboronic acid

12513-25G 12513-5G
76907-008EA 453.01 USD
76907-008 76907-010
3-(Propylcarbamoyl)phenylboronic acid ≥97%
3-(Propylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04038919
CAS Number: 850567-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR