3-(Methylsulphonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid

12460-25G 12460-5G
76906-992EA 215.37 USD
76906-992 76906-994
3-(Methylsulphonyl)phenylboronic acid ≥97%
3-(Methylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₄S
Storage Temperature: Ambient
MDL Number: MFCD03092935
CAS Number: 373384-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR