3-Chloro-2-fluoroisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-fluoropyridine-4-carboxylic acid

12394-1G 12394-25G 12394-500MG 12394-5G
76906-980EA 216.43 USD
76906-980 76906-982 76906-984 76906-986
3-Chloro-2-fluoroisonicotinic acid ≥97%
3-Chloro-2-fluoroisonicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031169
CAS Number: 741683-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR