4-(Pentyloxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amyloxyphenylboronic acid, 4-Pentyloxyphenylboronic acid, (4-Pentyloxyphenyl)boronic acid, 4-Pentyloxybenzeneboronic acid, 4-n-Pentyloxybenzeneboronic acid, 4-(N-Pentyloxy)phenylboronic acid, 4-Amyloxybenzeneboronic acid

12295-25G 12295-5G
76906-960EA 268.41 USD
76906-960 76906-962
4-(Pentyloxy)phenylboronic acid ≥97%
4-(Pentyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD04039035
CAS Number: 146449-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR