4-(4-(Trifluoromethoxy)phenoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

121888-1G 121888-25G 121888-5G
76906-932EA 107.16 USD
76906-932 76906-934 76906-936
4-(4-(Trifluoromethoxy)phenoxy)aniline ≥97%
4-(4-(Trifluoromethoxy)phenoxy)aniline
Formula: C₁₃H₁₀F₃NO₂
MW: 269.219 g/mol
Storage Temperature: Ambient
CAS Number: 58119-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR