(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4'-Isopropoxyphenyl)phenylboronic acid

12125-1G 12125-25G 12125-5G
76906-910EA 81.69 USD
76906-910 76906-912 76906-914
(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)boronic acid
Formula: C₁₅H₁₇BO₃
MW: 256.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411292
CAS Number: 870717-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR