4-Benzyloxy-3-formylbenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Formyl-4-benzyloxyphenylboronic acid, (4-(Benzyloxy)-3-formylphenyl)boronic acid

11946-1G 11946-25G 11946-5G
76906-862EA 94.43 USD
76906-862 76906-864 76906-866
4-Benzyloxy-3-formylbenzeneboronic acid ≥97%
4-Benzyloxy-3-formylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BO₄
MW: 256.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179487
CAS Number: 121124-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR