3,5-Dichloro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11875-1G 11875-25G 11875-5G
76906-838EA 109.28 USD
76906-838 76906-840 76906-842
3,5-Dichloro-4-methoxyphenylboronic acid ≥97%
3,5-Dichloro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04039018
CAS Number: 175883-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR