3,5-Diformylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3,5-Diformylphenyl)boronic acid

11867-1G 11867-25G 11867-500MG 11867-5G
76906-828EA 173.99 USD
76906-828 76906-830 76906-832 76906-834
3,5-Diformylphenylboronic acid ≥97%
3,5-Diformylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BO₄
MW: 177.9498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701570
CAS Number: 480424-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR