3,5-Dichloro-4-methoxybenzenecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dichloro-4-methoxybenzoic acid

11814-25G 11814-5G
76906-816EA 183.54 USD
76906-816 76906-818
3,5-Dichloro-4-methoxybenzenecarboxylic acid ≥97%
3,5-Dichloro-4-methoxybenzenecarboxylic acid
Formula: C₈H₆Cl₂O₃
MW: 221.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210404
CAS Number: 37908-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR