3-(Cyclopropylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Cyclopropylaminocarbonyl)phenylboronic acid

11442-25G 11442-5G
76906-744EA 381.93 USD
76906-744 76906-746
3-(Cyclopropylcarbamoyl)phenylboronic acid ≥97%
3-(Cyclopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04115694
CAS Number: 850567-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR