3-Chloro-4-propoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11430-25G 11430-5G
76906-736EA 275.84 USD
76906-736 76906-738
3-Chloro-4-propoxyphenylboronic acid ≥97%
3-Chloro-4-propoxyphenylboronic acid
Formula: C₉H₁₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD03701555
CAS Number: 480438-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR