4-Bromo-1-methyl-2-(methylsulfonyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-(methylsulphonyl)toluene

113468-25G 113468-5G
76906-708EA 584.56 USD
76906-708 76906-710
4-Bromo-1-methyl-2-(methylsulfonyl)benzene ≥97% research grade
4-Bromo-1-methyl-2-(methylsulfonyl)benzene
Formula: C₈H₉BrO₂S
MW: 249.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037925
CAS Number: 254887-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR