(3-Fluoro-4-hydroxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid

11226-25G 11226-5G
76906-672EA 527.27 USD
76906-672 76906-674
(3-Fluoro-4-hydroxyphenyl)boronic acid ≥97% research grade
(3-Fluoro-4-hydroxyphenyl)boronic acid
Formula: C₅H₅BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06659838
CAS Number: 182344-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR