3-Formyl-2-thiopheneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Formylthiophene-2-boronic acid, (3-Formylthiophen-2-yl)boronic acid

11220-25G 11220-5G
76906-666EA 453.01 USD
76906-666 76906-668
3-Formyl-2-thiopheneboronic acid ≥97%
3-Formyl-2-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075679
CAS Number: 17303-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR