3,5-Difluoro-2-nitrophenol ≥97%

Supplier: AOBChem USA

Warning

111216-25G 111216-5G
76906-638EA 346.92 USD
76906-638 76906-640
3,5-Difluoro-2-nitrophenol ≥97%
3,5-Difluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD08458165
CAS Number: 151414-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR