(3-Bromo-2-isopropoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-isopropoxyphenylboronic acid

11103-1G 11103-25G 11103-5G
76906-628EA 70.02 USD
76906-628 76906-630 76906-632
(3-Bromo-2-isopropoxyphenyl)boronic acid ≥97%
(3-Bromo-2-isopropoxyphenyl)boronic acid
Formula: C₉H₁₂BBrO₃
MW: 258.90 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664347
CAS Number: 870718-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR