4-Benzyloxy-3,5-dimethylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

11080-1G 11080-25G 11080-5G
76906-610EA 62.6 USD
76906-610 76906-612 76906-614
4-Benzyloxy-3,5-dimethylphenylboronic acid ≥97% research grade
4-Benzyloxy-3,5-dimethylphenylboronic acid
Formula: C₁₅H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD04039003
CAS Number: 333788-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR