4-Bromothiophene-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromothiophen-3-yl)boronic acid, 3-Bromothiophene-4-boronic acid

11009-25G 11009-5G
76906-580EA 552.73 USD
76906-580 76906-582
4-Bromothiophene-3-boronic acid ≥97%
4-Bromothiophene-3-boronic acid

Precaución: for Research Use Only

Formula: C₄H₄BBrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD06801663
CAS Number: 101084-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR