3-(tert-Butylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(tert-Butylaminocarbonyl)phenylboronic acid

10978-25G 10978-5G
76906-558EA 509.24 USD
76906-558 76906-560
3-(tert-Butylcarbamoyl)phenylboronic acid ≥97%
3-(tert-Butylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04115693
CAS Number: 183158-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR