(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-ethoxy-5-fluorophenylboronic acid

10842-25G 10842-5G
76906-510EA 630.18 USD
76906-510 76906-512
(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid ≥97% research grade
(3-Bromo-2-ethoxy-5-fluorophenyl)boronic acid
Formula: C₈H₉BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD05865158
CAS Number: 352534-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR